Ruthenium(III) chloride, anhydrous 99.9% (trace metals basis)

Supplier: Thermo Fisher Scientific
Danger

197210020 197210100 197210500
ACRO197210020EA 190 EUR
ACRO197210020 ACRO197210100 ACRO197210500
Ruthenium(III) chloride, anhydrous 99.9% (trace metals basis)
Ruthenium (III) chloride
Formula: Cl₃Ru
Melting Pt: >500 °C (dec.)
Density: 3,11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011208
CAS Number: 10049-08-8
EINECS: 233-167-5
UN: 3260
ADR: 8,II
Merck Index: 12,08455

Order Now

Specification Test Results

Appearance Black Powder or crystals
Assay 48 to 50 % (Ru)
Total trace metal impurities ≤0.1 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR