S-Methyl methanethiosulphonate (MMTS) 97%

Supplier: Thermo Fisher Scientific
Warning

197390010 197390050 197390250 197391000
ACRO197390010EA 59.9 EUR
ACRO197390010 ACRO197390050 ACRO197390250 ACRO197391000
S-Methyl methanethiosulphonate (MMTS) 97%
S-Methyl methanethiosulphonate (MMTS)

Appearance: Clear yellow to orange Liquid

Formula: C₂H₆O₂S₂
Boiling Pt: 69…71 °C (,4 mmHg)
Density: 1,227 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00007565
CAS Number: 2949-92-0
UN: 3334
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear yellow to orange Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR