Tungsten(VI) chloride 99% 20 mesh

Supplier: Thermo Fisher Scientific
Danger

200300100 200300500 200302500
ACRO200300100EA 75.5 EUR
ACRO200300100 ACRO200300500 ACRO200302500
Tungsten(VI) chloride 99% 20 mesh
Tungsten (VI) chloride
Formula: Cl₆W
Boiling Pt: 347 °C (760 mmHg)
Melting Pt: 275 °C
Density: 3,518 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011463
CAS Number: 13283-01-7
EINECS: 236-293-9
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Appearance Grey to dark blue to violet to black Powder
Purity ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR