Tetrachlorosilane 99.8+%

Supplier: Thermo Fisher Scientific
Danger

201160500 201165000
ACRO201160500EA 36.3 EUR
ACRO201160500 ACRO201165000
Tetrachlorosilane 99.8+%
Tetrachlorosilane
Formula: Cl₄Si
Boiling Pt: 58 °C (760 mmHg)
Melting Pt: –70 °C
Density: 1,48 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011229
CAS Number: 10026-04-7
EINECS: 233-054-0
UN: 1818
ADR: 8,II
Merck Index: 12,08644

Order Now

Specification Test Results

Appearance Clear colorless Liquid (May develop some turbidity or precipitate)
Purity ≥99.80 %
Aluminium (Al) ≤5 ppm
Antimony (Sb) ≤0.1 ppm
Chromium (Cr) ≤0.2 ppm
Cobalt (Co) ≤0.1 ppm
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤1 ppm
Manganese (Mn) ≤0.1 ppm
Molybdenum (Mo) ≤0.1 ppm
Nickel (Ni) ≤0.1 ppm
Titanium (Ti) ≤1 ppm
Vanadium (V) ≤0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR