Mercury 99.99+%

Supplier: Thermo Fisher Scientific
Danger

201421000 201425000 201420025
ACRO201421000EA 55.8 EUR
ACRO201421000 ACRO201425000 ACRO201420025
Mercury 99.99+%
Mercury
Formula: Hg
Boiling Pt: 356,5 °C (1013 hPa)
Melting Pt: –38,9 °C
Density: 13,54 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011035
CAS Number: 7439-97-6
EINECS: 231-106-7
UN: 2809
ADR: 8,III
Merck Index: 13,05925

Order Now

Specification Test Results

Appearance Silver white Metallic liquid
Assay ≥99.990 %
Cadmium (Cd) ≤0.1 ppm
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤0.5 ppm
Lead (Pb) ≤0.1 ppm
Silver (Ag) ≤1 ppm
Zinc (Zn) ≤0.5 ppm
Insoluble matter ≤0.002 % (in HNO3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR