Ammonium cerium(IV) nitrate 99% for analysis

Supplier: Thermo Fisher Scientific
Danger

201441000 201442500 201440010 201440050
ACRO201441000EA 101 EUR
ACRO201441000 ACRO201442500 ACRO201440010 ACRO201440050
Ammonium cerium(IV) nitrate 99% for analysis
Ammonium cerium (IV) nitrate
Formula: (NH₄)₂Ce(NO₃)₆
Melting Pt: 107…108 °C
Density: 2,2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151121
CAS Number: 16774-21-3
EINECS: 240-827-6
UN: 3085
ADR: 5.1,II
Merck Index: 13,00509

Order Now

Specification Test Results

Appearance Orange Crystalline powder or crystals
Solubility (10 % in water) Clear
Moisture ≤2 %
Titration ≥98.5 % (on dry substance)
Calcium oxide (CaO) ≤5 ppm
Iron(III)oxide (Fe2O3) ≤5 ppm
Potassium oxide (K2O) ≤3 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR