Hydrazinium monochloride 98+%

Supplier: Thermo Fisher Scientific
Danger

201990010 201992500 201990051
ACRO201990010EA 288 EUR
ACRO201990010 ACRO201992500 ACRO201990051
Hydrazinium monochloride 98+%
Hydrazinium monochloride
Formula: H₅ClN₂
Melting Pt: 89…93 °C
Density: 1,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00044368
CAS Number: 2644-70-4
EINECS: 220-154-4
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR