1-Fluoro-4-iodobenzene 99%

Supplier: Thermo Fisher Scientific
Warning

204500010 204500050 204500500
ACRO204500010EA 21.9 EUR
ACRO204500010 ACRO204500050 ACRO204500500
1-Fluoro-4-iodobenzene 99%
1-Fluoro-4-iodobenzene
Formula: C₆H₄FI
Boiling Pt: 182…184 °C (760 mmHg)
Melting Pt: –20 °C
Density: 1,925 g/cm³
Flash Pt: 68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001052
CAS Number: 352-34-1
EINECS: 206-522-7

Order Now

Specification Test Results

Appearance Clear yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5820 to 1.5840 (20°C, 589 nm)
GC ≥98.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR