Florisil® 100-200 mesh for column chromatography

Supplier: Thermo Fisher Scientific

205442500 205445000 205440050
ACRO205442500EA 164 EUR
ACRO205442500 ACRO205445000 ACRO205440050
Florisil® 100-200 mesh for column chromatography
Magnesium silicate
Formula: MgO₃Si
Storage Temperature: Ambient
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1

Order Now

Specification Test Results

Appearance White Powder
Particle size 7.8 % (325 mesh)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR