Manganese(II) acetate tetrahydrate 99+% for analysis

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Manganese acetate tetrahydrate, Manganese diacetate tetrahydrate, Acetic acid manganese (II) salt tetrahydrate

205660250 205665000 205660025
ACRO205660250EA 32.8 EUR
ACRO205660250 ACRO205665000 ACRO205660025
Manganese(II) acetate tetrahydrate 99+% for analysis
Manganese (II) acetate tetrahydrate
Formula: (H₃CCOO)₂Mn·4H₂O
Melting Pt: >300 °C
Density: 1,58 g/cm³ (20 °C)
Flash Pt: >130 °C
Storage Temperature: Ambient
MDL Number: MFCD00062552
CAS Number: 6156-78-1
EINECS: 211-334-3
Merck Index: 13,05746

Order Now

Specification Test Results

Appearance Light pink Crystals
Titration Complexometric ≥99.0 %
pH 6 to 7 (5 % solution)
Chloride (Cl) ≤20 ppm
Sulfate (SO4) ≤50 ppm
Copper (Cu) ≤5 ppm
Iron (Fe) ≤50 ppm
Nickel (Ni) ≤20 ppm
Insoluble matter ≤0.01 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR