Allylpalladium(II) chloride dimer 98%

Supplier: Thermo Fisher Scientific
Warning

206835000 206830050
ACRO206835000EA 116 EUR
ACRO206835000 ACRO206830050
Allylpalladium(II) chloride dimer 98%
Allylpalladium(II) chloride dimer

CAS No.: 12012-95-2

Formula: C₆H₁₀Cl₂Pd₂
Storage Temperature: Freezer
MDL Number: MFCD00044874
CAS Number: 12012-95-2

Order Now

Specification Test Results

Appearance Yellow to yellow-green Crystalline powder or crystals
Assay ≥56.7 % (Pd)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR