Bis(benzonitrile)palladium(II) dichloride 97+%

Supplier: Thermo Fisher Scientific
Warning

207900010 207900100
ACRO207900010EA 112 EUR
ACRO207900010 ACRO207900100
Bis(benzonitrile)palladium(II) dichloride 97+%
Bis(benzonitrile)dichloropalladium(II)
Formula: C₁₄H₁₀Cl₂N₂Pd
Melting Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00013123
CAS Number: 14220-64-5
EINECS: 238-085-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to orange to brown Powder
Infrared spectrum Conforms
Elemental Analysis ≥97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR