Methotrexate hydrate 99%

Supplier: Thermo Fisher Scientific
Danger

208100250 208101000 208105000
ACRO208100250EA 65.8 EUR
ACRO208100250 ACRO208101000 ACRO208105000
Methotrexate hydrate 99%
Methotrexate hydrate
Formula: C₂₀H₂₂N₈O₅·xH₂O
Melting Pt: 195 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00150847
CAS Number: 133073-73-1
EINECS: 200-413-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Water ≤13 % (K.F.)
HPLC ≥98.5 %
Specific optical rotation +19° to +24° (20°C, 589 nm) (c=1, 14 g/l Na2CO3) (on anhydrous substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR