Zirconyl chloride octahydrate 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Zirconium(IV) oxide chloride octahydrate, Zirconium dichloride oxide octahydrate, Zirconium(IV) oxychloride octahydrate

208370050 208371000 208375000
ACRO208370050EA 44.4 EUR
ACRO208370050 ACRO208371000 ACRO208375000
Zirconyl chloride octahydrate 98%
Zirconyl chloride octahydrate
Formula: Cl₂OZr·8H₂O
MW: 322.25 g/mol
Melting Pt: > 150 °C
Density: 1.91 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149898
CAS Number: 13520-92-8
EINECS: 231-717-9
UN: 3260
ADR: 8 II

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Titration Argentometric 97.5 to 102.5 %
Water 42 to 50 %
Solubility (10% in water) Clear to slightly hazy solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR