Sodium bromate 99+%, extra pure

Supplier: Thermo Fisher Scientific
Danger

208871000 208875000 208870025
ACRO208871000EA 36 EUR
ACRO208871000 ACRO208875000 ACRO208870025
Sodium bromate 99+%, extra pure
Sodium bromate
Formula: BrNaO₃
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 381 °C (dec.)
Density: 3,33 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003476
CAS Number: 7789-38-0
EINECS: 232-160-4
UN: 1494
ADR: 5.1,II
Merck Index: 12,08736

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Iodometric ≥99.0 %
Bromide (Br) ≤500 ppm
Sulfate (SO4) ≤500 ppm
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR