O-Methylhydroxylamine hydrochloride 98+%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Methylhydroxylamine hydrochloride, O-Methylhydroxylammonium chloride, Methoxyamine hydrochloride, Methoxylamine hydrochloride

210490050 210490250 210491000
ACRO210490050EA 113 EUR
ACRO210490050 ACRO210490250 ACRO210491000
O-Methylhydroxylamine hydrochloride 98+%
O-Methylhydroxylamine hydrochloride
Formula: CH₆ClNO
MW: 83.515 g/mol
Melting Pt: 148…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00012951
CAS Number: 593-56-6
EINECS: 209-798-7
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 148 to 153 °C
Titration with HClO4 98.0 to 102.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR