Hexane (mixture of isomers), technical

Supplier: Thermo Fisher Scientific
Danger

210590010 210590025 210590050 210590250
ACRO210590010EA 38.2 EUR
ACRO210590010 ACRO210590025 ACRO210590050 ACRO210590250
Hexane (mixture of isomers), technical
Hexane (mixture of isomers)
Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 69 °C
Melting Pt: –95 °C
Density: 0,659 g/cm³
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 92112-69-1
EINECS: 295-570-2
UN: 1208
ADR: 3,II
REACH: 01-2119484651-34
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Color scale ≤10 APHA
Residue after evaporation ≤0.002 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR