Benzyl isocyanate 99+%

Supplier: Thermo Fisher Scientific
Danger

210760050 210760250
ACRO210760050EA 75.9 EUR
ACRO210760050 ACRO210760250
Benzyl isocyanate 99+%
Benzyl isocyanate
Formula: C₈H₇NO
Boiling Pt: 101…104 °C (33 mmHg)
Density: 1,07 g/cm³
Flash Pt: 45 °C
MDL Number: MFCD00009701
CAS Number: 3173-56-6
EINECS: 221-640-9
UN: 3080
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5241 to 1.5261 (20°C, 589 nm)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR