Cerium trichloride heptahydrate 99%

Supplier: Thermo Fisher Scientific
Danger

211821000 211825000 211820250
ACRO211821000EA 108 EUR
ACRO211821000 ACRO211825000 ACRO211820250
Cerium trichloride heptahydrate 99%
Cerium trichloride heptahydrate
Formula: CeCl₃·7H₂O
Melting Pt: 848 °C
Density: 3,97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149634
CAS Number: 18618-55-8
EINECS: 232-227-8
UN: 1759
ADR: 8,II
Merck Index: 12,02041

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Titration Argentometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR