New fuchsine

Supplier: Thermo Fisher Scientific
Warning

212140250 212141000
ACRO212140250EA 47.4 EUR
ACRO212140250 ACRO212141000
New fuchsine
New fuchsine
Formula: C₂₂H₂₄ClN₃
Storage Temperature: Ambient
MDL Number: MFCD00012567
CAS Number: 3248-91-7
EINECS: 221-831-7

Order Now

Specification Test Results

Appearance Dark green Crystalline powder or crystals
Infrared spectrum Conforms
UV Lambda max 550 to 554 nm (in 50% ethanol)
Absorption Ratio P-15/P+15 1.1 to 1.4
Specific absorbance A (1%/1cm) ≥1500 (at lambda max.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR