Gallium trinitrate hydrate 99.9998% (trace metals basis)

Supplier: Thermo Fisher Scientific
Danger

212440010 212440050 212440250 212441000
ACRO212440010EA 52.8 EUR
ACRO212440010 ACRO212440050 ACRO212440250 ACRO212441000
Gallium trinitrate hydrate 99.9998% (trace metals basis)
Gallium trinitrate hydrate
Formula: GaN₃O₉·H₂O
Melting Pt: 110 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149723
CAS Number: 69365-72-6
EINECS: 236-815-5
UN: 1477
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Total trace metal impurities ≤0.0002 %
Gallium (Ga) 18 to 22 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR