Antimony(III) potassium oxitartrate trihydrate ACS

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (R,R)-(+)-Antimony potassium tartrate trihydrate, (+)-Antimony potassium L-tartrate trihydrate, Tartar emetic, Antimony potassium L(+)-tartrate trihydrate, L(+)-Tartaric acid antimony potassium salt trihydrate, Potassium Antimony (III) Oxide Tartrate Trihydrate, (R,R)-(+)-Tartaric acid antimony potassium salt trihydrate, L(+)-Antimony potassium tartrate trihydrate, Potassium antimony(III) oxide tartrate trihydrate, Antimony potassium (+) tartrate trihydrate, Potassium antimony tartrate trihydrate, Potassium antimonyl tartrate sesquihydrate, Antimony potassium tartrate trihydrate, Potassium antimonyl tartrate trihydrate

212801000
ACRO212801000EA 78.9 EUR
ACRO212801000
Antimony(III) potassium oxitartrate trihydrate ACS
Antimony (III) potassium oxitartrate trihydrate
Formula: C₈H₄K₂O₁₂Sb₂·3H₂O
Melting Pt: 100 °C
Density: 2,6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167056
CAS Number: 28300-74-5
UN: 1551
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Iodometric 99.0 to 103.0 %
Loss on drying ≤2.7 %
Arsenic (As) ≤0.015 %
Titrable acid or base ≤0.020 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR