5-Hexen-1-ol 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Hexen-6-ol, 5-Hexen-1-ol

213040050 213040250 213041000
ACRO213040050EA 99.2 EUR
ACRO213040050 ACRO213040250 ACRO213041000
5-Hexen-1-ol 99%
5-Hexen-1-ol
Formula: C₆H₁₂O
Boiling Pt: 152…155 °C (760 mmHg)
Melting Pt: -20 °C
Density: 0,8725 g/cm³
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00002981
CAS Number: 821-41-0
EINECS: 212-477-4
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4360 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR