Pyridinium-p-toluenesulphonate 98%

Supplier: Thermo Fisher Scientific
Warning

213740250 213740050 213741000 213745000
ACRO213740250EA 98.5 EUR
ACRO213740250 ACRO213740050 ACRO213741000 ACRO213745000
Pyridinium-p-toluenesulphonate 98%
Pyridinium p-toluenesulphonate
Formula: C₁₂H₁₃NO₃S
Melting Pt: 116…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00013108
CAS Number: 24057-28-1
EINECS: 246-002-7

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 112 to 119 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR