Cobalt(II) chloride, anhydrous 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Cobaltous chloride, Cobalt dichloride

214130050 214131000 214135000
ACRO214130050EA 57.3 EUR
ACRO214130050 ACRO214131000 ACRO214135000
Cobalt(II) chloride, anhydrous 97%
Cobalt (II) chloride
Formula: Cl₂Co
Boiling Pt: 1049 °C (1013 hPa)
Melting Pt: 735 °C
Density: 1,6795 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00010938
CAS Number: 7646-79-9
EINECS: 231-589-4
UN: 3077
ADR: 9,III
Merck Index: 12,02498

Order Now

Specification Test Results

Appearance Blue Powder
Titration Complexometric ≥96.0 %
Iron (Fe) ≤500 ppm
Lead (Pb) ≤100 ppm
Nickel (Ni) ≤2000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR