2,6-Bis(trifluoromethyl)benzoic acid 98%

Supplier: Thermo Fisher Scientific
Warning

214460010 214460050
ACRO214460010EA 40.7 EUR
ACRO214460010 ACRO214460050
2,6-Bis(trifluoromethyl)benzoic acid 98%
2,6-Bis(trifluoromethyl)benzoic acid
Formula: C₉H₄F₆O₂
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 137,7…138.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00000376
CAS Number: 24821-22-5

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 136 to 140 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR