Tributyltin hydride 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Tri-n-butyltin hydride

215730100 215730500 215732500
ACRO215730100EA 50.9 EUR
ACRO215730100 ACRO215730500 ACRO215732500
Tributyltin hydride 97%
Tributyltin hydride
Formula: C₁₂H₂₈Sn
Boiling Pt: 80 °C (,4 mmHg)
Melting Pt: ? 0 °C
Density: 1,08 g/cm³
Flash Pt: 40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009416
CAS Number: 688-73-3
EINECS: 211-704-4
UN: 2788
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4720 to 1.4740 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR