Methyl salicylate 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Wintergreen Oil, Methyl 2-hydroxybenzoate

220490010 220490050 220495000 220490025
ACRO220490010EA 121 EUR
ACRO220490010 ACRO220490050 ACRO220495000 ACRO220490025
Methyl salicylate 99%
Methyl salicylate
Formula: 2-(HO)C₆H₄CO₂CH₃
MW: 152.15 g/mol
Boiling Pt: 222 °C (760 mmHg)
Melting Pt: 8…7 °C
Density: 1.1825 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002214
CAS Number: 119-36-8
EINECS: 204-317-7
Merck Index: 13,06143
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d636eb895c055579d494c4b8ed1636c32f052dd7&vid=7f7da4bc95980831012b381705f4b5a3f4ab7003&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5350 to 1.5380 (20°C, 589 nm)
Density (20/4°C) 1.180 to 1.186 g/ml
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR