Tetrabutylammonium fluoride trihydrate 99%

Supplier: Thermo Fisher Scientific
Danger

221080100 221080500 221082500
ACRO221080100EA 52.4 EUR
ACRO221080100 ACRO221080500 ACRO221082500
Tetrabutylammonium fluoride trihydrate 99%
Tetrabutylammonium fluoride trihydrate
Formula: C₁₆H₃₆FN·3H₂O
Melting Pt: 58…60 °C
MDL Number: MFCD00149981
CAS Number: 87749-50-6
EINECS: 207-057-2
UN: 1759
ADR: 8,III

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR