Pyridinium chloride 98%, pure

Supplier: Thermo Fisher Scientific
Warning

222281000 222285000 222280025
ACRO222281000EA 51.3 EUR
ACRO222281000 ACRO222285000 ACRO222280025
Pyridinium chloride 98%, pure
Pyridinium chloride
Formula: C₅H₆ClN
Boiling Pt: 222,5 °C (1013 hPa)
Melting Pt: 140…146 °C
Density: 1,34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012802
CAS Number: 628-13-7
EINECS: 211-027-4

Order Now

Specification Test Results

Appearance White to tan Crystals
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
Solubility almost colorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR