Chromium(III) potassium sulphate dodecahydrate 98+%

Supplier: Thermo Fisher Scientific
Warning

222520050 222521000 222525000 222520025
ACRO222520050EA 38.3 EUR
ACRO222520050 ACRO222521000 ACRO222525000 ACRO222520025
Chromium(III) potassium sulphate dodecahydrate 98+%
Chromium (III) potassium sulphate dodecahydrate
Formula: CrK(SO₄)₂·12H₂O
Melting Pt: 89 °C
Density: 1,83 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149917
CAS Number: 7788-99-0
EINECS: 233-401-6
Merck Index: 13,02250

Order Now

Specification Test Results

Appearance Violet to dark purple Crystalline powder or crystals
Titration Iodometric 98.0 to 102.0 %
Chloride (Cl) ≤20 ppm
Aluminium (Al) ≤200 ppm
Ammonium (NH4) ≤100 ppm
Iron (Fe) ≤100 ppm
Insoluble matter ≤0.01 % (in water)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR