Nickel acetate tetrahydrate 99+% for analysis

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Nickel diacetate tetrahydrate, Nickel (II) acetate tetrahydrate, Nickel di(acetate) tetrahydrate, Acetic acid nickel (II) salt tetrahydrate

223145000 223140025 223141000
ACRO223145000EA 93.3 EUR
ACRO223145000 ACRO223140025 ACRO223141000
Nickel acetate tetrahydrate 99+% for analysis
Nickel acetate tetrahydrate
Formula: C₄H₆NiO₄·4H₂O
Boiling Pt: 16 °C (1013 hPa)
Melting Pt: 250 °C
Density: 1,798 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066973
CAS Number: 6018-89-9
EINECS: 206-761-7
UN: 3077
ADR: 9,III
Merck Index: 12,06583

Order Now

Specification Test Results

Appearance Green to blue Crystalline powder
Infrared spectrum Conforms
Titration Complexometric ≥99.0 %
Aluminium (Al) ≤5 ppm
Cadmium (Cd) ≤50 ppm
Calcium (Ca) ≤100 ppm
Chromium (Cr) ≤50 ppm
Cobalt (Co) ≤2000 ppm
Copper (Cu) ≤50 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm
Zinc (Zn) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR