Cetrimonium bromide 99+%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide

227160100 227160250 227161000 227165000 227160025 227160101
ACRO227160100EA 30.4 EUR
ACRO227160100 ACRO227160250 ACRO227161000 ACRO227165000 ACRO227160025 ACRO227160101
Cetrimonium bromide 99+%
Cetrimonium bromide
Formula: CH₃(CH₃)₁₅N(Br)(CH₃)₃
MW: 364.45 g/mol
Melting Pt: 230 °C
Flash Pt: 244 °C
Storage Temperature: Ambient
MDL Number: MFCD00011772
CAS Number: 57-09-0
EINECS: 200-311-3
UN: 3077
ADR: 9 III
Merck Index: 13,02034

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR