1-Naphthyl acetate 99%

Supplier: Thermo Fisher Scientific

227250250
ACRO227250250EA 90.2 EUR
ACRO227250250
1-Naphthyl acetate 99%
1-Naphthyl acetate
Formula: C₁₂H₁₀O₂
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 43…46 °C
Density: 1,1305 g/cm³ (50 °C)
Flash Pt: ≥ 100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003922
CAS Number: 830-81-9
EINECS: 212-599-8

Order Now

Specification Test Results

Appearance White to light yellow to light pink Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 42 to 47 °C
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR