Methyl viologen dichloride hydrate 98%

Supplier: Thermo Fisher Scientific
Danger

227320010 227320050
ACRO227320010EA 92.8 EUR
ACRO227320010 ACRO227320050
Methyl viologen dichloride hydrate 98%
Methyl viologen dichloride hydrate

CAS No.: 1910-42-5

Formula: C₁₂H₁₄Cl₂N₂·nH₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150001
CAS Number: 1910-42-5
UN: 2811
ADR: 6.1,I

Order Now

Specification Test Results

Appearance Beige to pink to brown Crystalline powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥97.5 % (on dry substance)
Water ≤18 % (K.F.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR