DL-Selenomethionine 99+%

Supplier: Thermo Fisher Scientific
Danger

240770010 240770025 240775000
ACRO240770010EA 320 EUR
ACRO240770010 ACRO240770025 ACRO240775000
DL-Selenomethionine 99+%
DL-Selenomethionine

Appearance: Colorless Glistening powder

Formula: C₅H₁₁NO₂Se
Melting Pt: 265 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063089
CAS Number: 1464-42-2
UN: 3283
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Colorless to white to off-white Glistening powder or flakes
Infrared spectrum Conforms
HPLC ≥99.0 %
Specific optical rotation -0.5° to +0.5° (20°C, 589 nm) (c=1, 1 N HCl)
Titration with HClO4 99.0 to 101.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR