5-Fluoro-2-nitrophenol 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Fluoro-2-hydroxynitrobenzene

241970250 241971000
ACRO241970250EA 79.3 EUR
ACRO241970250 ACRO241971000
5-Fluoro-2-nitrophenol 99%
5-Fluoro-2-nitrophenol
Formula: C₆H₄FNO₃
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 32…35 °C
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00007107
CAS Number: 446-36-6
EINECS: 207-168-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to orange-brown Powder or crystals or crystalline mass and/or chunks
Infrared spectrum Conforms
Melting point 32 to 35 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR