Fmoc-Osu (N-(9-fluorenylmethoxycarbonyloxy)succinimide) 98%

Supplier: Thermo Fisher Scientific
Warning

243100050 243100250
ACRO243100050EA 82.8 EUR
ACRO243100050 ACRO243100250
Fmoc-Osu (N-(9-fluorenylmethoxycarbonyloxy)succinimide) 98%
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimide)
Formula: C₁₉H₁₅NO₅
Melting Pt: 145…153 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010733
CAS Number: 82911-69-1
EINECS: 433-520-5
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 145 to 153 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR