2,5-Dihydrofuran 98%

Supplier: Thermo Fisher Scientific
Danger

244780250 244781000
ACRO244780250EA 73.7 EUR
ACRO244780250 ACRO244781000
2,5-Dihydrofuran 98%
2,5-Dihydrofuran
Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 66…67 °C (760mbar)
Density: 0.92 g/cm³
Flash Pt: –016 °C
Storage Temperature: Ambient
MDL Number: MFCD00003219
CAS Number: 1708-29-8
UN: 3271
ADR: 3 II

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4310 to 1.4330 (20°C, 589 nm)
GC ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR