(±)-trans-1,2-Diaminocyclohexane 98%

Supplier: Thermo Fisher Scientific
Danger

248670100 248670500 248672500
ACRO248670100EA 37.5 EUR
ACRO248670100 ACRO248670500 ACRO248672500
(±)-trans-1,2-Diaminocyclohexane 98%
(±)-trans-1,2-Diaminocyclohexane
Formula: C₆H₁₄N₂
Boiling Pt: 79…81 °C (15 mmHg)
Melting Pt: 14…15 °C
Density: 0,951 g/cm³
Flash Pt: 68 °C
MDL Number: MFCD00063747
CAS Number: 1121-22-8
UN: 2735

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid (May develop some turbidity or precipitate)
Infrared spectrum Conforms
Refractive index 1.4870 to 1.4900 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR