L(+)-Ribose 99%

Supplier: Thermo Fisher Scientific

Synonyms: (2S,3S,4S)-2,3,4,5-Tetrahydroxypentanal, L-(+)-Ribose

257910010 257910050 257910250
ACRO257910010EA 41.3 EUR
ACRO257910010 ACRO257910050 ACRO257910250
L(+)-Ribose 99%
L(+)-Ribose
Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Boiling Pt: 331
Melting Pt: 84…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00167010
CAS Number: 24259-59-4

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 85 to 92 °C
Water ≤0.5 %
Solubility (10 % in water) Clear colorless
HPLC ≥98.5 % (ELSD)
Specific optical rotation +19° to +21° (20 °C, 589 nm) (c=2, H2O)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR