(±)-1-Ethoxy-2-propanol 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Ethoxy-2-propanol, (±)-Monopropylene glycol ethyl ether

259050010 259050025 259050100
ACRO259050010EA 72 EUR
ACRO259050010 ACRO259050025 ACRO259050100
(±)-1-Ethoxy-2-propanol 95%
(±)-1-Ethoxy-2-propanol
Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 132 °C (760 mmHg)
Melting Pt: –100 °C
Density: 0.897 g/cm³
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00067050
CAS Number: 1569-02-4
EINECS: 216-374-5
UN: 3271
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4050 to 1.4090 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥94 % Remark: balance 2-ethoxy-1-propanol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR