2-(2-(Piperazin-1-yl)ethoxy)ethanol 95%

Supplier: Thermo Fisher Scientific

259110050 259110250 259111000
ACRO259110050EA 56.8 EUR
ACRO259110050 ACRO259110250 ACRO259111000
2-(2-(Piperazin-1-yl)ethoxy)ethanol 95%
2-(2-(Piperazin-1-yl)ethoxy)ethanol

CAS No.: 13349-82-1

Formula: C₈H₁₈N₂O₂
Boiling Pt: 150 °C (13 mmHg)
Density: 1,01 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00074931
CAS Number: 13349-82-1

Order Now

Specification Test Results

Appearance Clear colorless to yellow-brown Viscous liquid
Infrared spectrum Conforms
Titration Tertiary base ≥94 %
GC ≥94 %
Titration with HClO4 ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR