2-Phenyl-5-benzimidazolesulphonic acid 98%

Supplier: Thermo Fisher Scientific

259700250 259700050 259702500
ACRO259700250EA 50.5 EUR
ACRO259700250 ACRO259700050 ACRO259702500
2-Phenyl-5-benzimidazolesulphonic acid 98%
2-Phenyl-5-benzimidazolesulphonic acid

CAS No.: 27503-81-7

Formula: C₁₃H₁₀N₂O₃S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00053007
CAS Number: 27503-81-7

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤7 %
Titration excess NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR