Oleic acid 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (9Z)-9-Octadecenoic acid

270290050 270290250 270291000
ACRO270290050EA 152 EUR
ACRO270290050 ACRO270290250 ACRO270291000
Oleic acid 97%
Oleic acid
Formula: C₁₈H₃₄O₂
Boiling Pt: 360 °C (760 mmHg)
Melting Pt: 13 °C
Density: 0,89 g/cm³
Flash Pt: 189 °C
Storage Temperature: Freezer
MDL Number: MFCD00064242
CAS Number: 112-80-1
EINECS: 204-007-1
Merck Index: 13,06898
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5aecc5dce4d0318386a45d7adb7c5822631010a&vid=892a2df4ecab3679b06983b84cd27ad334ef961f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4580 to 1.4610 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR