2-Ethylhexylamine 99%

Supplier: Thermo Fisher Scientific
Danger

271660010 271662500 271660025 271661000
ACRO271660010EA 101 EUR
ACRO271660010 ACRO271662500 ACRO271660025 ACRO271661000
2-Ethylhexylamine 99%
2-Ethylhexylamine
Formula: C₈H₁₉N
Boiling Pt: 169 °C (760 mmHg)
Melting Pt: –76 °C
Density: 0,78 g/cm³
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00008148
CAS Number: 104-75-6
EINECS: 203-233-8
UN: 2276
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4290 to 1.4320 (20°C, 589 nm)
Water ≤0.2 %
GC ≥98.5 %
Color scale ≤20 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR