(S)-(+)-Camptothecin 98%

Supplier: Thermo Fisher Scientific
Danger

276721000 276720010
ACRO276721000EA 279 EUR
ACRO276721000 ACRO276720010
(S)-(+)-Camptothecin 98%
(S)-(+)-Camptothecin

CAS No.: 7689-03-4

Formula: C₂₀H₁₆N₂O₄
Melting Pt: 260 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00081076
CAS Number: 7689-03-4
UN: 1544
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Loss on drying ≤1.0 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR