(Aminomethyl)phosphonic acid 99%

Supplier: Thermo Fisher Scientific
Danger

291640010 291642500 291640050
ACRO291640010EA 170 EUR
ACRO291640010 ACRO291642500 ACRO291640050
(Aminomethyl)phosphonic acid 99%
(Aminomethyl)phosphonic acid
Formula: CH₆NO₃P
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008105
CAS Number: 1066-51-9
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR