4,4'-Di-tert-butylbiphenyl 99+%

Supplier: Thermo Fisher Scientific

291880050 291880250
ACRO291880050EA 61.5 EUR
ACRO291880050 ACRO291880250
4,4'-Di-tert-butylbiphenyl 99+%
4,4'-Di-tert-butylbiphenyl
Formula: C₂₀H₂₆
Boiling Pt: 190…192 °C (13 mmHg)
Melting Pt: 126…129 °C
Density: 0,9691 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008834
CAS Number: 1625-91-8
EINECS: 216-615-4

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 126 to 129 °C
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR