3-Cyclohexene-1-carboxylic acid 97%

Supplier: Thermo Fisher Scientific
Danger

294390100 294390010
ACRO294390100EA 220 EUR
ACRO294390100 ACRO294390010
3-Cyclohexene-1-carboxylic acid 97%
3-Cyclohexene-1-carboxylic acid

Appearance: Clear yellow-brown to blue-grey Liquid

Formula: C₇H₁₀O₂
Boiling Pt: 238 °C (760 mmHg)
Melting Pt: 17 °C
Density: 1,08 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00013781
CAS Number: 4771-80-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear yellow-brown to blue-grey Liquid
Infrared spectrum Conforms
Refractive index 1.4800 to 1.4820 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR